trans-2-(tert-Butoxycarbonyl)-5-hydroxy-2-azabicyclo[2.2.1]heptane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

115557-250MG 115557-500MG 115557-1G
102896-836EA 2226.15 USD
102896-836 102896-838 102896-840
trans-2-(tert-Butoxycarbonyl)-5-hydroxy-2-azabicyclo[2.2.1]heptane-3-carboxylic acid ≥95%
trans-2-(tert-Butoxycarbonyl)-5-hydroxy-2-azabicyclo[2.2.1]heptane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₉NO₅
MDL Number: MFCD20231322
CAS Number: 1250884-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR