1-Tosylazetidin-3-one ≥95%

Supplier: Matrix Scientific

115556-100MG 115556-250MG 115556-500MG
102896-830EA 989.23 USD
102896-830 102896-832 102896-834
1-Tosylazetidin-3-one ≥95%
1-Tosylazetidin-3-one

Hazard: Irritant

Formula: C₁₀H₁₁NO₃S
MDL Number: MFCD00129860
CAS Number: 76543-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR