Octahydropyrazino[2,3-d]azepine-1,7-dicarboxylic acid 7-benzylester 1-tert-butyl ester ≥95%

Supplier: Matrix Scientific

115554-250MG 115554-500MG 115554-1G
102896-818EA 1647.69 USD
102896-818 102896-820 102896-822
Octahydropyrazino[2,3-d]azepine-1,7-dicarboxylic acid 7-benzylester 1-tert-butyl ester ≥95%
Octahydropyrazino[2,3-d]azepine-1,7-dicarboxylic acid 7-benzylester 1-tert-butyl ester

Hazard: Irritant

Formula: C₂₁H₃₁N₃O₄
MDL Number: MFCD14581300
CAS Number: 1251004-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR