Octahydroisoindole-2,5-dicarboxylic acid 2-tert-butyl ester ≥95%

Supplier: Matrix Scientific

115553-250MG 115553-500MG 115553-1G
102896-812EA 1373.85 USD
102896-812 102896-814 102896-816
Octahydroisoindole-2,5-dicarboxylic acid 2-tert-butyl ester ≥95%
Octahydroisoindole-2,5-dicarboxylic acid 2-tert-butyl ester

Hazard: Irritant

Formula: C₁₄H₂₃NO₄
MW: 269.34 g/mol
MDL Number: MFCD17016445
CAS Number: 1250885-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR