3-Methyl-5-(tributylstannyl)pyridine ≥97%

Supplier: Matrix Scientific

115551-500MG 115551-1G 115551-5G
102899-440EA 344.62 USD
102899-440 102899-442 102899-444
3-Methyl-5-(tributylstannyl)pyridine ≥97%
3-Methyl-5-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₈H₃₃NSn
MDL Number: MFCD22056726
CAS Number: 167556-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR