Methyl-2-(((benzyloxy)carbonyl)amino)-2-(oxetan-3-ylidene)acetate ≥95%

Supplier: Matrix Scientific

115550-1G 115550-5G 115550-10G
102896-806EA 989.23 USD
102896-806 102899-436 102899-438
Methyl-2-(((benzyloxy)carbonyl)amino)-2-(oxetan-3-ylidene)acetate ≥95%
Methyl-2-(((benzyloxy)carbonyl)amino)-2-(oxetan-3-ylidene)acetate

Hazard: Irritant

Formula: C₁₄H₁₅NO₅
MDL Number: MFCD14586429
CAS Number: 394653-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR