Ethyl (2-nitro-5-(pentafluorosulfanyl)phenyl) acetate ≥95%
Supplier: Matrix Scientific
115548-500MG
115548-1G
102896-798EA
1044.62
USD
102896-798
102896-800
Ethyl (2-nitro-5-(pentafluorosulfanyl)phenyl) acetate ≥95%
Ethyl (2-nitro-5-(pentafluorosulfanyl)phenyl) acetate
Hazard: Irritant
Formula:
C₁₀H₁₀F₅NO₄S MW: 335.25 g/mol |
MDL Number:
MFCD22370090 CAS Number: 1309569-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...