3-(1,1-Dioxo-1-thiomorpholine-4-yl)azetidine-1-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

115547-100MG 115547-250MG 115547-500MG
102896-792EA 413.85 USD
102896-792 102896-794 102896-796
3-(1,1-Dioxo-1-thiomorpholine-4-yl)azetidine-1-carboxylic acid tert-butyl ester ≥95%
3-(1,1-Dioxo-1-thiomorpholine-4-yl)azetidine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₄S
MW: 290.38 g/mol
MDL Number: MFCD22370084
CAS Number: 1257293-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR