3-Chloro-4-(tributylstannyl)pyridine ≥96%
Supplier: Matrix Scientific
115545-1G
115545-5G
115545-25G
102896-780EA
413.85
USD
102896-780
102896-782
102896-784
3-Chloro-4-(tributylstannyl)pyridine ≥96%
3-Chloro-4-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₇H₃₀ClNSn |
MDL Number:
MFCD22370076 CAS Number: 444989-46-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...