[(1-Chloro-2-methylcyclopropyl)sulfinyl]benzene ≥95%

Supplier: Matrix Scientific

115544-100MG 115544-250MG
102896-776EA 1318.46 USD
102896-776 102896-778
[(1-Chloro-2-methylcyclopropyl)sulfinyl]benzene ≥95%
[(1-Chloro-2-methylcyclopropyl)sulfinyl]benzene

Hazard: Irritant

Formula: C₁₀H₁₁ClOS
MDL Number: MFCD22370073
CAS Number: 1004968-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR