[(1-Chloro-2,3-dimethylcyclopropyl)thio]benzene ≥95%

Supplier: Matrix Scientific

115543-100MG 115543-250MG
102896-772EA 1044.62 USD
102896-772 102896-774
[(1-Chloro-2,3-dimethylcyclopropyl)thio]benzene ≥95%
[(1-Chloro-2,3-dimethylcyclopropyl)thio]benzene

Hazard: Irritant

Formula: C₁₁H₁₃ClS
MDL Number: MFCD22370072
CAS Number: 64474-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR