[(1-Chloro-2,3-dimethylcyclopropyl)sulfinyl]benzene ≥95%

Supplier: Matrix Scientific

115541-100MG 115541-250MG
102896-764EA 1318.46 USD
102896-764 102896-766
[(1-Chloro-2,3-dimethylcyclopropyl)sulfinyl]benzene ≥95%
[(1-Chloro-2,3-dimethylcyclopropyl)sulfinyl]benzene

Hazard: Irritant

Formula: C₁₁H₁₃ClOS
MDL Number: MFCD22370070
CAS Number: 65006-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR