[(1-Chloro-2,2-dimethylcyclopropyl)sulfinylbenzene ≥95%

Supplier: Matrix Scientific

115540-100MG 115540-250MG
102896-760EA 1318.46 USD
102896-760 102896-762
[(1-Chloro-2,2-dimethylcyclopropyl)sulfinylbenzene ≥95%
[(1-Chloro-2,2-dimethylcyclopropyl)sulfinylbenzene

Hazard: Irritant

Formula: C₁₁H₁₃ClOS
MDL Number: MFCD22370069
CAS Number: 65006-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR