[(1-Chlorocyclopropyl)thio]benzene ≥95%

Supplier: Matrix Scientific

115539-100MG 115539-250MG 115539-500MG
102896-754EA 715.38 USD
102896-754 102896-756 102896-758
[(1-Chlorocyclopropyl)thio]benzene ≥95%
[(1-Chlorocyclopropyl)thio]benzene

Hazard: Irritant

Formula: C₉H₉ClS
MDL Number: MFCD22370068
CAS Number: 64416-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR