[(1-Chlorocyclopropyl)sulfinyl]benzene ≥98%

Supplier: Matrix Scientific

Synonyms: ((1-Chlorocyclopropyl)sulphinyl)benzene

115538-100MG 115538-250MG 115538-500MG
102896-748EA 604.62 USD
102896-748 102896-750 102896-752
[(1-Chlorocyclopropyl)sulfinyl]benzene ≥98%
[(1-Chlorocyclopropyl)sulfinyl]benzene

Hazard: Irritant

Formula: C₉H₉ClOS
MW: 200.69 g/mol
MDL Number: MFCD22370067
CAS Number: 65006-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR