tert-Butyl-4-(1H-pyrrolo[2,3-b]pyridin-2-yl)piperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
115535-500MG
115535-1G
115535-5G
102896-732EA
1512.31
USD
102896-732
102896-734
102896-736
tert-Butyl-4-(1H-pyrrolo[2,3-b]pyridin-2-yl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(1H-pyrrolo[2,3-b]pyridin-2-yl)piperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₇H₂₃N₃O₂ |
MDL Number:
MFCD12912636 CAS Number: 1198283-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...