cis-tert-Butyl-8-oxo-3-azabicyclo-[4.2.0]octane-3-carboxylate ≥95%

Supplier: Matrix Scientific

115534-250MG 115534-500MG 115534-1G
102896-726EA 1373.85 USD
102896-726 102896-728 102896-730
cis-tert-Butyl-8-oxo-3-azabicyclo-[4.2.0]octane-3-carboxylate ≥95%
cis-tert-Butyl-8-oxo-3-azabicyclo-[4.2.0]octane-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₉NO₃
MDL Number: MFCD12198720
CAS Number: 1251004-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR