tert-Butyl-3-(nitromethylene)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(nitromethylene)azetidine-1-carboxylate

115533-250MG 115533-500MG
102896-722EA 1867.69 USD
102896-722 102896-724
tert-Butyl-3-(nitromethylene)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(nitromethylene)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₉H₁₄N₂O₄
MW: 214.22 g/mol
MDL Number: MFCD20486581
CAS Number: 1379812-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR