tert-Butyl-3-hydroxy-1-oxa-6-azaspiro[3.3]heptane-6-carboxylate ≥95%

Supplier: Matrix Scientific

115530-250MG 115530-500MG 115530-1G
102896-650EA 1593.85 USD
102896-650 102896-652 102896-654
tert-Butyl-3-hydroxy-1-oxa-6-azaspiro[3.3]heptane-6-carboxylate ≥95%
tert-Butyl-3-hydroxy-1-oxa-6-azaspiro[3.3]heptane-6-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
MDL Number: MFCD22370059
CAS Number: 1349199-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR