(3aS*,6aS*)-tert-Butylhexahydropyrrolo-[3,4-c]pyrrole-2(1H)-carboxylate ≥95%

Supplier: Matrix Scientific

115529-250MG 115529-500MG 115529-1G
102896-530EA 1373.85 USD
102896-530 102896-532 102896-534
(3aS*,6aS*)-tert-Butylhexahydropyrrolo-[3,4-c]pyrrole-2(1H)-carboxylate ≥95%
(3aS*,6aS*)-tert-Butylhexahydropyrrolo-[3,4-c]pyrrole-2(1H)-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₉NO₅
MDL Number: MFCD12198700
CAS Number: 1251019-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR