1-tert-Butyl-4-ethyl-4-(4-fluorobenzyl)piperidine-1,4-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-tert-Butyl 4-ethyl 4-(4-fluorobenzyl)piperidine-1,4-dicarboxylate

115528-500MG 115528-1G 115528-5G
102896-524EA 536.92 USD
102896-524 102896-526 102896-528
1-tert-Butyl-4-ethyl-4-(4-fluorobenzyl)piperidine-1,4-dicarboxylate ≥95%
1-tert-Butyl-4-ethyl-4-(4-fluorobenzyl)piperidine-1,4-dicarboxylate

Hazard: Irritant

Formula: C₂₀H₂₈FNO₄
MW: 365.45 g/mol
MDL Number: MFCD09038430
CAS Number: 917755-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR