tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl bis(2-hydroxyethyl)carbamate, N-BOC-Diethanolamine

115526-1G 115526-5G
102896-640EA 249.23 USD
102896-640 102896-642
tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate ≥97%
tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate

Hazard: Irritant

Formula: C₉H₁₉NO₄
MW: 205.26 g/mol
MDL Number: MFCD01862953
CAS Number: 103898-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR