tert-Butyl-5-(benzyloxycarbonylamino)-2-azabicyclo[2.1.1]hexane-2-carboxylate ≥95%

Supplier: Matrix Scientific

115525-250MG 115525-500MG 115525-1G
102896-634EA 1647.69 USD
102896-634 102896-636 102896-638
tert-Butyl-5-(benzyloxycarbonylamino)-2-azabicyclo[2.1.1]hexane-2-carboxylate ≥95%
tert-Butyl-5-(benzyloxycarbonylamino)-2-azabicyclo[2.1.1]hexane-2-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₄
MW: 332.4 g/mol
MDL Number: MFCD12198746
CAS Number: 1250885-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR