tert-Butyl (1S,5R)-6-amino-3-azabicyclo[3.2.0]heptane-3-carboxylate ≥95%

Supplier: Matrix Scientific

115524-250MG 115524-500MG 115524-1G
102896-628EA 1647.69 USD
102896-628 102896-630 102896-632
tert-Butyl (1S,5R)-6-amino-3-azabicyclo[3.2.0]heptane-3-carboxylate ≥95%
tert-Butyl (1S,5R)-6-amino-3-azabicyclo[3.2.0]heptane-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
MDL Number: MFCD14581158
CAS Number: 1250884-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR