(3aR*,8R*,8aR*)-tert-Butyl-8-amino-3,3a,8,8a-tetra-hydroindeno[2,1-c]pyrrole-2(1H)-carboxylate ≥95%

Supplier: Matrix Scientific

115523-250MG 115523-500MG 115523-1G
102896-622EA 1373.85 USD
102896-622 102896-624 102896-626
(3aR*,8R*,8aR*)-tert-Butyl-8-amino-3,3a,8,8a-tetra-hydroindeno[2,1-c]pyrrole-2(1H)-carboxylate ≥95%
(3aR*,8R*,8aR*)-tert-Butyl-8-amino-3,3a,8,8a-tetra-hydroindeno[2,1-c]pyrrole-2(1H)-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₂
MDL Number: MFCD14581080
CAS Number: 1251007-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR