1'-(tert-Butoxycarbonyl)spiro[bicyclo[2.2.1]hept[2]ene-7,4'-piperidine]-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

115522-250MG 115522-500MG 115522-1G
102899-430EA 1647.69 USD
102899-430 102899-432 102899-434
1'-(tert-Butoxycarbonyl)spiro[bicyclo[2.2.1]hept[2]ene-7,4'-piperidine]-5-carboxylic acid ≥95%
1'-(tert-Butoxycarbonyl)spiro[bicyclo[2.2.1]hept[2]ene-7,4'-piperidine]-5-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₅NO₄
MDL Number: MFCD12198776
CAS Number: 1251015-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR