(1S*,3aR*,4R*,6aS*)-2-Benzyl 1-ethyl-4-aminohexahydrocyclopenta[c]pyrrole-1,2(1H)-dicarboxylate ≥95%

Supplier: Matrix Scientific

115516-250MG 115516-500MG 115516-1G
102896-592EA 1647.69 USD
102896-592 102896-594 102896-596
(1S*,3aR*,4R*,6aS*)-2-Benzyl 1-ethyl-4-aminohexahydrocyclopenta[c]pyrrole-1,2(1H)-dicarboxylate ≥95%
(1S*,3aR*,4R*,6aS*)-2-Benzyl 1-ethyl-4-aminohexahydrocyclopenta[c]pyrrole-1,2(1H)-dicarboxylate

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₄
MDL Number: MFCD12198731
CAS Number: 1251021-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR