Benzyl-3,6-diazabicyclo[3.2.2]nonane-6-carboxylate ≥95%

Supplier: Matrix Scientific

115515-250MG 115515-500MG 115515-1G
102896-586EA 1647.69 USD
102896-586 102896-588 102896-590
Benzyl-3,6-diazabicyclo[3.2.2]nonane-6-carboxylate ≥95%
Benzyl-3,6-diazabicyclo[3.2.2]nonane-6-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₂
MW: 260.34 g/mol
MDL Number: MFCD17016711
CAS Number: 1251013-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR