2-Azabicyclo[2.2.1]heptane-2,7-dicarboxylic acid-2-tert-butyl ester ≥95%

Supplier: Matrix Scientific

115510-250MG 115510-500MG 115510-1G
102896-558EA 1373.85 USD
102896-558 102896-560 102896-562
2-Azabicyclo[2.2.1]heptane-2,7-dicarboxylic acid-2-tert-butyl ester ≥95%
2-Azabicyclo[2.2.1]heptane-2,7-dicarboxylic acid-2-tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₁₉NO₄
MDL Number: MFCD17016716
CAS Number: 1251007-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR