4-(Piperidin-4-ylsulfanyl)-pyridine dihydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
4-[(Piperidin-4-yl)thio]pyridine dihydrochloride
115505-250MG
115505-500MG
115505-1G
102896-520EA
769.23
USD
102896-520
102896-522
102896-536
4-(Piperidin-4-ylsulfanyl)-pyridine dihydrochloride ≥95%
4-(Piperidin-4-ylsulfanyl)-pyridine dihydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₆Cl₂N₂S MW: 267.22 g/mol |
MDL Number:
MFCD03840146 CAS Number: 105283-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...