Ethyl-1-benzyl-5,5-difluoro-1,2,5,6-tetrahydropyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

115504-250MG 115504-500MG 115504-1G
102896-406EA 1787.69 USD
102896-406 102896-408 102896-518
Ethyl-1-benzyl-5,5-difluoro-1,2,5,6-tetrahydropyridine-3-carboxylate ≥95%
Ethyl-1-benzyl-5,5-difluoro-1,2,5,6-tetrahydropyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₇F₂NO₂
MDL Number: MFCD22199428
CAS Number: 1373503-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR