Ethyl-2-(1-benzyl-3,3-difluoropiperidin-4-ylidene)acetate ≥95%

Supplier: Matrix Scientific

115503-250MG 115503-500MG 115503-1G
102896-400EA 1593.85 USD
102896-400 102896-402 102896-404
Ethyl-2-(1-benzyl-3,3-difluoropiperidin-4-ylidene)acetate ≥95%
Ethyl-2-(1-benzyl-3,3-difluoropiperidin-4-ylidene)acetate

Hazard: Irritant

Formula: C₁₆H₁₉F₂NO₂
MDL Number: MFCD22199402
CAS Number: 1373510-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR