1-tert-Butyl-4-ethyl-5-fluoro-4-methyl-3,4-dihydropyridine-1,4(2H)-dicarboxylate ≥95%

Supplier: Matrix Scientific

115499-1G 115499-250MG 115499-500MG
102896-380EA 2666.15 USD
102896-380 102896-514 102896-516
1-tert-Butyl-4-ethyl-5-fluoro-4-methyl-3,4-dihydropyridine-1,4(2H)-dicarboxylate ≥95%
1-tert-Butyl-4-ethyl-5-fluoro-4-methyl-3,4-dihydropyridine-1,4(2H)-dicarboxylate

Hazard: Irritant

Formula: C₁₄H₂₂FNO₄
MDL Number: MFCD22199427
CAS Number: 1373503-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR