1-tert-Butyl-3-ethyl-5,5-difluoropiperidine-1,3-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-tert-Butyl 3-ethyl 5,5-difluoropiperidine-1,3-dicarboxylate

115498-250MG 115498-500MG 115498-1G
102896-508EA 1593.85 USD
102896-508 102896-510 102896-512
1-tert-Butyl-3-ethyl-5,5-difluoropiperidine-1,3-dicarboxylate ≥95%
1-tert-Butyl-3-ethyl-5,5-difluoropiperidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₂₁F₂NO₄
MW: 293.31 g/mol
MDL Number: MFCD21601880
CAS Number: 1356339-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR