1-Boc-3-trifluoromethylpiperidin-4-one ≥95%

Supplier: Matrix Scientific

115495-250MG 115495-500MG 115495-1G
102899-410EA 3241.54 USD
102899-410 102899-412 102899-414
1-Boc-3-trifluoromethylpiperidin-4-one ≥95%
1-Boc-3-trifluoromethylpiperidin-4-one

Hazard: Irritant

Formula: C₁₁H₁₆F₃NO₃
MDL Number: MFCD08669340
CAS Number: 1159983-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR