Benzo[1,2-b:4,5-b']dithiophene ≥98%

Supplier: Matrix Scientific

115493-500MG 115493-1G 115493-5G
102891-292EA 344.62 USD
102891-292 102891-294 102891-296
Benzo[1,2-b:4,5-b']dithiophene ≥98%
Benzo[1,2-b:4,5-b']dithiophene

Hazard: Irritant

Formula: C₁₀H₆S₂
CAS Number: 267-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR