Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde ≥96%

Supplier: Matrix Scientific

Synonyms: Thieno[3,2-b]thiophene-2,5-dicarbaldehyde

115490-500MG 115490-1G
102896-502EA 1318.46 USD
102896-502 102896-504
Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde ≥96%
Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde

Hazard: Irritant

Formula: C₈H₄O₂S₂
MW: 196.24 g/mol
MDL Number: MFCD03931117
CAS Number: 37882-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR