2,4-cis-Piperidine-2,4-dicarboxylic acid ≥95%
Supplier: Matrix Scientific
115489-500MG
115489-1G
115489-5G
102896-496EA
1098.46
USD
102896-496
102896-498
102896-500
2,4-cis-Piperidine-2,4-dicarboxylic acid ≥95%
2,4-cis-Piperidine-2,4-dicarboxylic acid
Hazard: Irritant
Formula:
C₇H₁₁NO₄ |
MDL Number:
MFCD00057330 CAS Number: 84211-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...