7-Methoxy-7-oxoheptanoic acid ≥95%

Supplier: Matrix Scientific

115488-1G 115488-5G
102896-490EA 549.23 USD
102896-490 102896-492
7-Methoxy-7-oxoheptanoic acid ≥95%
7-Methoxy-7-oxoheptanoic acid

Hazard: Irritant

Formula: C₈H₁₄O₄
MW: 174.2 g/mol
MDL Number: MFCD00040445
CAS Number: 20291-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR