Ethyl-1-benzyl-4-hydroxypiperidine-3-carboxylate ≥95%
Supplier: Matrix Scientific
115486-500MG
115486-1G
115486-5G
102896-478EA
360
USD
102896-478
102896-480
102896-482
Ethyl-1-benzyl-4-hydroxypiperidine-3-carboxylate ≥95%
Ethyl-1-benzyl-4-hydroxypiperidine-3-carboxylate
Hazard: Irritant
Formula:
C₁₅H₂₁NO₃ |
MDL Number:
MFCD19687152 CAS Number: 956010-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...