Ethyl-1-benzyl-3-hydroxypiperidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

115485-500MG 115485-1G 115485-5G
102896-472EA 360 USD
102896-472 102896-474 102896-476
Ethyl-1-benzyl-3-hydroxypiperidine-4-carboxylate ≥95%
Ethyl-1-benzyl-3-hydroxypiperidine-4-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₁NO₃
MDL Number: MFCD21602034
CAS Number: 850040-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR