2,2,2-Trifluoro-1-(piperazin-1-yl)ethanone hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2,2-Trifluoro-1-(piperazin-1-yl)ethanonehydrochloride

115484-1G 115484-5G
102896-468EA 275.38 USD
102896-468 102896-470
2,2,2-Trifluoro-1-(piperazin-1-yl)ethanone hydrochloride ≥95%
2,2,2-Trifluoro-1-(piperazin-1-yl)ethanone hydrochloride

Hazard: Irritant

Formula: C₆H₁₀ClF₃N₂O
MW: 218.61 g/mol
MDL Number: MFCD22421756
CAS Number: 245487-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR