tert-Butyl-4-(2,2,2-trifluoroacetyl)piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115483-1G 115483-5G
102896-464EA 275.38 USD
102896-464 102896-466
tert-Butyl-4-(2,2,2-trifluoroacetyl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(2,2,2-trifluoroacetyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₇F₃N₂O₃
MW: 282.26 g/mol
MDL Number: MFCD22421755
CAS Number: 77278-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR