1-(tert-Butoxycarbonyl)-4,4-difluoropiperidine-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
115480-250MG
115480-500MG
115480-1G
102896-454EA
4393.85
USD
102896-454
102896-456
102896-458
1-(tert-Butoxycarbonyl)-4,4-difluoropiperidine-3-carboxylic acid ≥95%
1-(tert-Butoxycarbonyl)-4,4-difluoropiperidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₇F₂NO₄ |
MDL Number:
MFCD18909829 CAS Number: 1303974-65-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...