1,7-Diazaspiro[4.5]decane-1-carboxylic acid benzyl ester ≥95%

Supplier: Matrix Scientific

115474-500MG 115474-1G
102896-432EA 2416.92 USD
102896-432 102896-434
1,7-Diazaspiro[4.5]decane-1-carboxylic acid benzyl ester ≥95%
1,7-Diazaspiro[4.5]decane-1-carboxylic acid benzyl ester

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₂
MDL Number: MFCD11223524
CAS Number: 1086394-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR