6-Boc-6-azaspiro[3.4]octane-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
6-(tert-Butoxycarbonyl)-6-azaspiro[3.4]octane-2-carboxylic acid
115471-500MG
115471-1G
102896-420EA
3515.38
USD
102896-420
102896-422
6-Boc-6-azaspiro[3.4]octane-2-carboxylic acid ≥95%
6-Boc-6-azaspiro[3.4]octane-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₂₁NO₄ MW: 255.32 g/mol |
MDL Number:
MFCD17016204 CAS Number: 1251002-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...