2-Amino-7-methyl-7-azaspiro[3.5]nonane ≥95%
Supplier: Matrix Scientific
115467-500MG
115467-1G
102896-368EA
3515.38
USD
102896-368
102896-370
2-Amino-7-methyl-7-azaspiro[3.5]nonane ≥95%
2-Amino-7-methyl-7-azaspiro[3.5]nonane
Hazard: Irritant
Formula:
C₉H₁₈N₂ MW: 154.26 g/mol |
MDL Number:
MFCD12198555 CAS Number: 1160247-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...