tert-Butyl-4-hydroxy-3-(trifluoromethyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-hydroxy-3-(trifluoromethyl)piperidine-1-carboxylate

115465-250MG 115465-500MG 115465-1G
102896-356EA 2953.85 USD
102896-356 102896-358 102896-360
tert-Butyl-4-hydroxy-3-(trifluoromethyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-hydroxy-3-(trifluoromethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₈F₃NO₃
MW: 269.27 g/mol
MDL Number: MFCD18800734
CAS Number: 1283720-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR