tert-Butyl-4-fluoro-3-(hydroxymethyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115463-250MG 115463-500MG 115463-1G
102896-344EA 2472.31 USD
102896-344 102896-346 102896-348
tert-Butyl-4-fluoro-3-(hydroxymethyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-fluoro-3-(hydroxymethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₀FNO₃
MDL Number: MFCD18909817
CAS Number: 1303972-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR