tert-Butyl-3-(benzylamino)-3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(benzylamino)-3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate

115461-100MG 115461-250MG
102899-388EA 1044.62 USD
102899-388 102899-390
tert-Butyl-3-(benzylamino)-3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(benzylamino)-3-(2-ethoxy-2-oxoethyl)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₉H₂₈N₂O₄
MW: 348.45 g/mol
MDL Number: MFCD22199157
CAS Number: 1373923-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR